Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PHT4;6
Entrez Gene ID
834463
Gene Name
phosphate transporter 4;6
Gene Aliases
AT5G44370, K9L2.16, K9L2_16, phosphate transporter 4;6
Chromosome Location
Chr.5: 17874734 - 17877658 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_123804.4 NP_199250.1 2 - 2 1927 137
GenBank mRNA BT002878.1 - 2 - 2 1763 137

View Full Product Details