Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LCR43
Entrez Gene ID
3768865
Gene Name
low-molecular-weight cysteine-rich 43
Gene Aliases
AT3G07005, low-molecular-weight cysteine-rich 43
Chromosome Location
Chr.3: 2212063 - 2212742 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035576.2 NP_001030653.1 1 - 2 68 110

View Full Product Details