Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TOPP2
Entrez Gene ID
836034
Gene Name
type one serine/threonine protein phosphatase 2
Gene Aliases
AT5G59160, MNC17.9, MNC17_9, PPO, PROTOPORPHYRINOGEN OXIDASE, type one serine/threonine protein phosphatase 2
Chromosome Location
Chr.5: 23879236 - 23881840 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001037026.1 NP_001032103.1 3 - 4 1187 75

View Full Product Details