Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
OCT4
Entrez Gene ID
821613
Gene Name
organic cation/carnitine transporter4
Gene Aliases
AT3G20660, AtOCT4, organic cation/carnitine transporter4
Chromosome Location
Chr.3: 7225037 - 7228797 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_112957.3 NP_188702.2 1 - 2 698 92
GenBank mRNA AK176424.1 - 1 - 2 496 92
BT010744.1 - 1 - 2 411 92

View Full Product Details