Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DTX1
Entrez Gene ID
814938
Gene Name
MATE efflux family protein
Gene Aliases
AT2G04040, ATDTX1, F3L12.13, F3L12_13, TX1, detoxification 1
Chromosome Location
Chr.2: 1334270 - 1336550 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_126443.5 NP_178491.1 3 - 4 955 143
GenBank mRNA AK227153.1 - 3 - 4 887 143
BT022022.1 - 3 - 4 894 143
BT026371.1 - 3 - 4 885 143

View Full Product Details