Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GF14 PHI
Entrez Gene ID
840403
Gene Name
GF14 protein phi chain
Gene Aliases
14-3-3 PROTEIN G-BOX FACTOR14 PHI, 14-3-3PHI, AT1G35160, GENERAL REGULATORY FACTOR 4, GF14 protein phi chain, GRF4, T32G9.30, T32G9_30
Chromosome Location
Chr.1: 12867081 - 12868890 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_103213.3 NP_564453.1 3 - 4 919 111
GenBank mRNA AY084342.1 - 3 - 4 819 111
L09111.1 - 3 - 4 787 111

View Full Product Details