Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BPC7
Entrez Gene ID
818120
Gene Name
basic pentacysteine 7
Gene Aliases
AT2G35550, ATBPC7, BASIC PENTACYSTEINE 7, BBR, T32F12.7, T32F12_7, basic pentacysteine 7
Chromosome Location
Chr.2: 14929243 - 14930898 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001036411.2 NP_001031488.1 2 - 2 867 89
NM_001084540.2 NP_001078009.1 2 - 2 838 89
NM_129107.3 NP_181098.2 2 - 2 949 89
NM_179918.2 NP_850249.1 2 - 2 791 89
GenBank mRNA AY102550.1 - 2 - 2 713 89
AY116857.1 - 2 - 2 924 89
AY380573.1 - 2 - 2 736 89
BT026350.1 - 1 - 1 584 89

View Full Product Details