Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RALFL6
Entrez Gene ID
5007821
Gene Name
RALF-like 6
Gene Aliases
AT1G60625, RALF-like 6
Chromosome Location
Chr.1: 22333916 - 22334161 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001084275.1 NP_001077744.1 1 - 1 149 102

View Full Product Details