Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RALFL19
Entrez Gene ID
817944
Gene Name
ralf-like 19
Gene Aliases
AT2G33775, ralf-like 19
Chromosome Location
Chr.2: 14288323 - 14289313 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_179888.4 NP_850219.1 1 - 1 846 158

View Full Product Details