The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001085329.1 | NP_001078798.1 | 2 - 3 | 365 | 137 |
NM_001126026.2 | NP_001119498.1 | 2 - 3 | 400 | 137 | |
NM_001345662.1 | NP_001332140.1 | 2 - 3 | 400 | 137 | |
NM_125904.3 | NP_569005.2 | 2 - 3 | 400 | 137 | |
GenBank mRNA | AF312667.1 | - | 2 - 3 | 376 | 137 |
AY141211.1 | - | 2 - 3 | 239 | 137 | |
AY231441.1 | - | 2 - 3 | 400 | 137 |