Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PRDM12
Entrez Gene ID
618320
Gene Name
PR/SET domain 12
Gene Aliases
-
Chromosome Location
Chr.11: 100951292 - 100966190 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206295.1 NP_001193224.1 2 - 3 419 76

View Full Product Details