Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TRIAP1
Entrez Gene ID
525164
Gene Name
TP53 regulated inhibitor of apoptosis 1
Gene Aliases
-
Chromosome Location
Chr.17: 62747153 - 62749467 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079776.1 NP_001073244.1 1 - 2 183 84
GenBank mRNA BC120172.1 - 1 - 2 183 84

View Full Product Details