Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PTGES3
Entrez Gene ID
493638
Gene Name
prostaglandin E synthase 3
Gene Aliases
CPGES
Chromosome Location
Chr.5: 56760782 - 56782195 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007806.2 NP_001007807.2 5 - 6 574 151
GenBank mRNA AY692440.1 - 4 - 5 378 151
BC103350.1 - 5 - 6 574 151

View Full Product Details