Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ANGPTL7
Entrez Gene ID
512698
Gene Name
angiopoietin like 7
Gene Aliases
-
Chromosome Location
Chr.16: 42447039 - 42453123 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014909.1 NP_001014909.1 4 - 5 1296 82
GenBank mRNA BT020703.1 - 4 - 5 1296 82
EU599230.1 - 4 - 5 869 82

View Full Product Details