Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
USP18
Entrez Gene ID
515202
Gene Name
ubiquitin specific peptidase 18
Gene Aliases
bubp43
Chromosome Location
Chr.5: 76001952 - 76018301 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017940.1 NP_001017940.1 7 - 8 867 64
GenBank mRNA AJ865078.1 - 7 - 8 867 64

View Full Product Details