Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RPS20
Entrez Gene ID
513222
Gene Name
ribosomal protein S20
Gene Aliases
-
Chromosome Location
Chr.14: 23278327 - 23279575 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034438.1 NP_001029610.1 1 - 1 90 104
GenBank mRNA BC103289.1 - 1 - 1 90 104

View Full Product Details