Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RCAN1
Entrez Gene ID
539640
Gene Name
regulator of calcineurin 1
Gene Aliases
DSCR1
Chromosome Location
Chr.1: 882073 - 1002231 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034679.1 NP_001029851.1 1 - 2 166 120
GenBank mRNA BC103185.1 - 1 - 2 166 120
BT030515.1 - 1 - 2 146 120

View Full Product Details