Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ELAVL1
Entrez Gene ID
617316
Gene Name
ELAV like RNA binding protein 1
Gene Aliases
HuR
Chromosome Location
Chr.7: 16715067 - 16752222 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076454.2 NP_001069922.1 2 - 3 346 108
GenBank mRNA BC102495.1 - 2 - 3 347 108

View Full Product Details