Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DCAKD
Entrez Gene ID
508778
Gene Name
dephospho-CoA kinase domain containing
Gene Aliases
-
Chromosome Location
Chr.19: 44717128 - 44737384 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077881.1 NP_001071349.1 2 - 3 258 83
GenBank mRNA BC103143.1 - 2 - 3 258 83

View Full Product Details