Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CPSF4
Entrez Gene ID
280875
Gene Name
cleavage and polyadenylation specific factor 4
Gene Aliases
NAR
Chromosome Location
Chr.25: 36959398 - 36971751 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_173942.2 NP_776367.1 1 - 2 239 130
GenBank mRNA U96448.1 - 1 - 2 239 130

View Full Product Details