Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PTH
Entrez Gene ID
280903
Gene Name
parathyroid hormone
Gene Aliases
-
Chromosome Location
Chr.15: 39190473 - 39193281 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_173954.2 NP_776379.2 2 - 3 216 80
GenBank mRNA J00024.1 - 2 - 3 181 80
M25082.1 - 2 - 3 216 80
V00106.1 - - 191 80

View Full Product Details