Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
APOE
Entrez Gene ID
281004
Gene Name
apolipoprotein E
Gene Aliases
-
Chromosome Location
Chr.18: 52635556 - 52640577 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_173991.2 NP_776416.1 1 - 2 40 66
GenBank mRNA BC102620.1 - 1 - 2 52 66
X64839.1 - - 40 66

View Full Product Details