Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CHAD
Entrez Gene ID
281069
Gene Name
chondroadherin
Gene Aliases
-
Chromosome Location
Chr.19: 36246817 - 36259301 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174019.2 NP_776444.1 1 - 2 819 65
GenBank mRNA GQ179990.1 - 1 - 2 750 65
U08018.1 - 1 - 2 819 65

View Full Product Details