Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CCNB2
Entrez Gene ID
281668
Gene Name
cyclin B2
Gene Aliases
-
Chromosome Location
Chr.10: 51184636 - 51209368 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174264.3 NP_776689.2 1 - 2 174 119
GenBank mRNA BC118382.1 - 1 - 2 81 119
BT020648.1 - 1 - 2 174 119
BT020669.1 - 1 - 2 146 119
BT020762.1 - 1 - 2 123 119
BT020822.1 - 1 - 2 126 119
BT021153.1 - 1 - 2 134 119
BT021700.1 - 1 - 2 123 119

View Full Product Details