Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CORO1A
Entrez Gene ID
282196
Gene Name
coronin 1A
Gene Aliases
-
Chromosome Location
Chr.25: 26134793 - 26140217 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174521.2 NP_776946.1 6 - 7 851 65
GenBank mRNA BC102876.1 - 6 - 7 920 65
BT025463.1 - 6 - 7 860 65
D44496.1 - 6 - 7 851 65

View Full Product Details