Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PAFAH1B2
Entrez Gene ID
282514
Gene Name
platelet activating factor acetylhydrolase 1b catalytic subunit 2
Gene Aliases
-
Chromosome Location
Chr.15: 27831374 - 27858164 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174664.2 NP_777089.1 4 - 5 704 155
GenBank mRNA AB098963.1 - 4 - 5 377 155
BC103452.1 - 4 - 5 390 155
D49678.1 - 4 - 5 704 155

View Full Product Details