Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ATP5MC1
Entrez Gene ID
338053
Gene Name
ATP synthase membrane subunit c locus 1
Gene Aliases
ATP5G1
Chromosome Location
Chr.19: 37618774 - 37621538 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_176649.3 NP_788822.1 1 - 2 82 103
GenBank mRNA X05218.1 - - 83 103

View Full Product Details