Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PPP2CA
Entrez Gene ID
282320
Gene Name
protein phosphatase 2 catalytic subunit alpha
Gene Aliases
-
Chromosome Location
Chr.7: 45843018 - 45867789 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_181031.2 NP_851374.1 6 - 7 986 85
GenBank mRNA BC147979.1 - 6 - 7 1075 85
BT021727.1 - 6 - 7 865 85
M16968.1 - 6 - 7 964 84
X52554.1 - - 986 85
X72858.1 - - 1024 85

View Full Product Details