Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NDUFA9
Entrez Gene ID
404188
Gene Name
NADH:ubiquinone oxidoreductase subunit A9
Gene Aliases
-
Chromosome Location
Chr.5: 105474046 - 105495864 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_205817.1 NP_991386.1 4 - 5 426 92
GenBank mRNA BC102359.1 - 4 - 5 428 92
BT025401.1 - 4 - 5 380 92
X59418.1 - - 426 92

View Full Product Details