Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
UPK3B
Entrez Gene ID
282115
Gene Name
uroplakin 3B
Gene Aliases
-
Chromosome Location
Chr.25: 34501342 - 34509115 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012672.1 NP_001012690.1 3 - 4 614 72
GenBank mRNA AY233461.1 - 3 - 4 614 72

View Full Product Details