Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IFT20
Entrez Gene ID
445364
Gene Name
intraflagellar transport 20
Gene Aliases
-
Chromosome Location
Chr.19: 19814951 - 19821939 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013603.1 NP_001013621.1 3 - 4 457 77
GenBank mRNA BC103453.1 - 3 - 4 484 77
BT021871.1 - 3 - 4 457 77

View Full Product Details