Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IFT20
Entrez Gene ID
445364
Gene Name
intraflagellar transport 20
Gene Aliases
-
Chromosome Location
Chr.19: 19814951 - 19821939 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013603.1 NP_001013621.1 5 - 6 651 149
GenBank mRNA BC103453.1 - 5 - 6 678 149
BT021871.1 - 5 - 6 651 149

View Full Product Details