Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC52A3
Entrez Gene ID
507459
Gene Name
solute carrier family 52 member 3
Gene Aliases
C13H20orf54, C20orf54, RFT2
Chromosome Location
Chr.13: 60290424 - 60307559 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014864.1 NP_001014864.1 3 - 4 1240 62
GenBank mRNA BC102546.1 - 3 - 4 1174 62
BT020859.1 - 3 - 4 1240 62

View Full Product Details