Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MOCS2
Entrez Gene ID
507986
Gene Name
molybdenum cofactor synthesis 2
Gene Aliases
-
Chromosome Location
Chr.20: 25956790 - 25975148 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014867.1 NP_001014867.1 3 - 4 322 147
GenBank mRNA BC123458.1 - 3 - 4 325 147
BT021515.1 - 3 - 4 322 147

View Full Product Details