Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DXO
Entrez Gene ID
508460
Gene Name
decapping exoribonuclease
Gene Aliases
DOM3Z
Chromosome Location
Chr.23: 27396745 - 27403613 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014872.2 NP_001014872.2 7 - 8 1388 85
GenBank mRNA BT021779.1 - 7 - 8 1388 85

View Full Product Details