Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MTG2
Entrez Gene ID
508880
Gene Name
mitochondrial ribosome associated GTPase 2
Gene Aliases
GTPBP5
Chromosome Location
Chr.13: 55018169 - 55025639 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014874.1 NP_001014874.1 1 - 2 38 89
GenBank mRNA BT021695.1 - 1 - 2 38 89

View Full Product Details