Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
JADE1
Entrez Gene ID
509264
Gene Name
jade family PHD finger 1
Gene Aliases
PHF17
Chromosome Location
Chr.17: 28943823 - 29000308 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001015559.1 NP_001015559.1 2 - 3 130 61
GenBank mRNA BT020833.1 - 2 - 3 130 61

View Full Product Details