Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PIGM
Entrez Gene ID
509680
Gene Name
phosphatidylinositol glycan anchor biosynthesis class M
Gene Aliases
-
Chromosome Location
Chr.3: 9644360 - 9647791 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001015563.1 NP_001015563.1 1 - 1 3252 93
GenBank mRNA BC150135.1 - 1 - 1 3205 93
BT020759.1 - 1 - 1 3252 93

View Full Product Details