Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DSTN
Entrez Gene ID
513267
Gene Name
destrin, actin depolymerizing factor
Gene Aliases
-
Chromosome Location
Chr.13: 37954770 - 37979552 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001015586.1 NP_001015586.1 3 - 4 440 108
GenBank mRNA BC103074.1 - 3 - 4 467 108
BT020985.1 - 3 - 4 440 108

View Full Product Details