Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CLPB
Entrez Gene ID
520639
Gene Name
caseinolytic mitochondrial matrix peptidase chaperone subunit B
Gene Aliases
SKD3
Chromosome Location
Chr.15: 51877179 - 52023017 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001015625.1 NP_001015625.1 12 - 13 1505 91
GenBank mRNA BC103469.1 - 12 - 13 1503 91
BT020885.1 - 12 - 13 1505 91

View Full Product Details