Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RRNAD1
Entrez Gene ID
532204
Gene Name
ribosomal RNA adenine dimethylase domain containing 1
Gene Aliases
C1orf66, C3H1orf66, CGI-41
Chromosome Location
Chr.3: 14098948 - 14105919 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001015641.1 NP_001015641.1 3 - 4 687 68
GenBank mRNA BC112638.1 - 3 - 4 582 68
BT020816.1 - 3 - 4 687 68

View Full Product Details