Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ZGPAT
Entrez Gene ID
510866
Gene Name
zinc finger CCCH-type and G-patch domain containing
Gene Aliases
-
Chromosome Location
Chr.13: 54037421 - 54047311 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024514.2 NP_001019685.2 5 - 6 1126 67
GenBank mRNA BC118131.1 - 5 - 6 1118 67
BT021717.1 - 4 - 5 1207 67

View Full Product Details