Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BCO1
Entrez Gene ID
534696
Gene Name
beta-carotene oxygenase 1
Gene Aliases
BCMO1, betaCO
Chromosome Location
Chr.18: 7897583 - 7928494 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024559.1 NP_001019730.1 10 - 11 1539 105
GenBank mRNA DQ008469.1 - 10 - 11 1539 105

View Full Product Details