Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
POLI
Entrez Gene ID
515909
Gene Name
DNA polymerase iota
Gene Aliases
-
Chromosome Location
Chr.24: 53691858 - 53717856 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024692.1 NP_001019863.1 4 - 5 589 78
GenBank mRNA AY965877.1 - 4 - 5 589 78
BC142234.1 - 4 - 5 469 78

View Full Product Details