Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ECHS1
Entrez Gene ID
281748
Gene Name
enoyl-CoA hydratase, short chain 1
Gene Aliases
-
Chromosome Location
Chr.26: 25629604 - 25638760 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025206.2 NP_001020377.2 7 - 8 834 72
GenBank mRNA BC109605.1 - 7 - 8 829 72
BT021569.1 - 7 - 8 834 72

View Full Product Details