Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PDPN
Entrez Gene ID
509732
Gene Name
podoplanin
Gene Aliases
-
Chromosome Location
Chr.16: 54010801 - 54043029 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033120.1 NP_001028292.1 2 - 3 187 120
GenBank mRNA AB205159.1 - 2 - 3 187 120
BC149398.1 - 2 - 3 390 120

View Full Product Details