Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
C7H1orf35
Entrez Gene ID
506656
Gene Name
chromosome 7 C1orf35 homolog
Gene Aliases
C1orf35, MMTAG2
Chromosome Location
Chr.7: 2998019 - 3000275 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034272.1 NP_001029444.1 4 - 5 431 81
GenBank mRNA BC120382.1 - 2 - 3 675 81
BT021507.1 - 4 - 5 431 81

View Full Product Details