Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IFI47
Entrez Gene ID
521987
Gene Name
interferon gamma inducible protein 47
Gene Aliases
-
Chromosome Location
Chr.7: 40459453 - 40475731 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034545.2 NP_001029717.1 1 - 2 197 104
GenBank mRNA BC102181.1 - 1 - 2 198 104

View Full Product Details