Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AAR2
Entrez Gene ID
533351
Gene Name
AAR2 splicing factor
Gene Aliases
C13H20orf4, C20orf4
Chromosome Location
Chr.13: 65359806 - 65380073 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034587.1 NP_001029759.1 1 - 2 314 108
GenBank mRNA BT021814.1 - 1 - 2 314 108

View Full Product Details