Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TNFRSF9
Entrez Gene ID
520341
Gene Name
TNF receptor superfamily member 9
Gene Aliases
-
Chromosome Location
Chr.16: 45439420 - 45457708 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035336.2 NP_001030413.1 5 - 6 550 90
GenBank mRNA BC102869.1 - 5 - 6 551 90

View Full Product Details